Novus Biologicals
Manufacturer Code:NBP29454620UL
Catalog # NB023617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human RRAGA (NP_006561.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenovirus E3 14.7 kDa-interacting protein 1; Adenovirus E3 14.7 kDa-interacting protein 1 FIP1 FIP-1adenovirus E3-14.7K interacting protein 1 Rag A RagA RAGAras-related GTP-binding protein A Ras-related GTP binding A; adenovirus E3-14.7K interacting protein 1; FIP-1; Rag A; Ras-related GTP binding A; Ras-related GTP-binding protein A; RRAGA
Gene Aliases: FIP-1; FIP1; RAGA; RRAGA
UniProt ID: (Human) Q7L523
Entrez Gene ID: (Human) 10670
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.