Invitrogen
Manufacturer Code:PA598696
Catalog # PIPA598696
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human CCR4-NOT transcription complex subunit 9 protein (1-258AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer/testis antigen 129; CCR4-NOT transcription complex subunit 9; cell differentiation protein RCD1 homolog; Cell differentiation protein RQCD1 homolog; EPO-induced protein FL10; protein involved in sexual development; Rcd-1; RCD1 required for cell differentiation1 homolog
Gene Aliases: 2610007F23Rik; AI593551; CAF40; CNOT9; CT129; Fl10; RCD-1; RCD1; RQCD1
UniProt ID: (Human) Q92600, (Mouse) Q9JKY0
Entrez Gene ID: (Human) 9125, (Mouse) 58184
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.