Invitrogen
Manufacturer Code:PA545811
Catalog # PIPA545811
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human RQCD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer/testis antigen 129; CCR4-NOT transcription complex subunit 9; cell differentiation protein RCD1 homolog; Cell differentiation protein RQCD1 homolog; EPO-induced protein FL10; protein involved in sexual development; Rcd-1; rcd1 (required for cell differentiation) homolog 1; RCD1 required for cell differentiation1 homolog; RCD1 required for cell differentiation1 homolog (S. pombe) CNOT9 CT129 RCD1 RCD1+
Gene Aliases: 2610007F23Rik; AI593551; CAF40; CNOT9; CT129; Fl10; RCD-1; RCD1; RQCD1; si:dkeyp-84a8.6; wu:fb52b11; wu:fb74e08; zgc:85618
UniProt ID: (Bovine) A7MB47, (Human) Q92600, (Mouse) Q9JKY0, (Rat) Q5PQL2, (Zebrafish) Q6NWL4
Entrez Gene ID: (Bovine) 536537, (Rabbit) 100351154, (Goat) 102176836, (Dog) 610449, (Human) 9125, (Horse) 100058624, (Guinea pig) 100714731, (Mouse) 58184, (Rat) 301513, (Zebrafish) 406632
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.