Invitrogen
Manufacturer Code:PA5106373
Catalog # PIPA5106373
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RQCD1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610007F23Rik AI593551 CAF40 cancer/testis antigen 129 CCR4-NOT transcription complex subunit 9 cell differentiation protein RCD1 homolog Cell differentiation protein RQCD1 homolog CNOT9 CT129 EPO-induced protein FL10 Fl10 protein involved in sexual development Rcd1 rcd-1 rcd1 (required for cell differentiation) homolog 1 rcd1 (required for cell differentiation) homolog 1 (S. pombe) RCD1 required for cell differentiation1 homolog Rcd1 required for cell differentiation1 homolog (S. pombe) Rqcd1; cancer/testis antigen 129; CCR4-NOT transcription complex subunit 9; cell differentiation protein RCD1 homolog; Cell differentiation protein RQCD1 homolog; EPO-induced protein FL10; protein involved in sexual development; Rcd-1; rcd1 (required for cell differentiation) homolog 1; RCD1 required for cell differentiation1 homolog
Gene Aliases: 2610007F23Rik; AI593551; CAF40; CNOT9; CT129; Fl10; RCD-1; RCD1; RQCD1
UniProt ID: (Human) Q92600, (Mouse) Q9JKY0, (Rat) Q5PQL2
Entrez Gene ID: (Human) 9125, (Mouse) 58184, (Rat) 301513
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.