Novus Biologicals
Manufacturer Code:NBP2834770.1ML
Catalog # NB031406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human RQCD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer/testis antigen 129; cancer/testis antigen 129 CNOT9protein involved in sexual development CT129 rcd-1 rcd1 (required for cell differentiation S.pombe) homolog 1 RCD1 required for cell differentiation1 homolog (S. pombe) RCD1+ RCD1cell differentiation protein RCD1 homolog; CCR4-NOT transcription complex subunit 9; Cell differentiation protein RQCD1 homolog; protein involved in sexual development; Rcd-1; RCD1 required for cell differentiation1 homolog
Gene Aliases: CAF40; CNOT9; CT129; RCD-1; RCD1; RQCD1
UniProt ID: (Human) Q92600
Entrez Gene ID: (Human) 9125
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.