Proteintech
Manufacturer Code:278431AP150UL
Catalog # 50-173-5773
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPUSD4 Fusion Protein Ag27225 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondrial RNA pseudouridine synthase RPUSD4; RNA pseudouridylate synthase domain-containing protein 4; RPUSD4
Gene Aliases: 2410001E19Rik; RGD1311268; RPUSD4
UniProt ID: (Human) Q96CM3, (Rat) Q4QQT0, (Mouse) Q9CWX4
Entrez Gene ID: (Human) 84881, (Rat) 315550, (Mouse) 71989
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.