Novus Biologicals
Manufacturer Code:NBP237297
Catalog # NBP237297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6G9C4 |
Immunogen | Purified recombinant fragment of human RPTOR (AA: 874-1009) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: KIAA1303p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats KOG1 Mip1 p150 target of rapamycin (TOR)-scaffold protein Raptor regulatory associated protein of MTOR complex 1 regulatory-associated protein of mTOR; p150 target of rapamycin (TOR)-scaffold protein; p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats; Raptor; regulatory associated protein of MTOR, complex 1; Regulatory-associated protein of mTOR
Gene Aliases: KIAA1303; KOG1; Mip1; RAPTOR; RPTOR
UniProt ID: (Human) Q8N122
Entrez Gene ID: (Human) 57521
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.