Abnova
Manufacturer Code:MAB9943
Catalog # 89-113-901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 33 |
Immunogen | Recombinant protein corresponding to full length human RPSA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against full-length recombinant RPSA.
For Research Use Only
Protein Aliases: 37 kDa laminin receptor; 37 kDa laminin receptor precursor; 37/67 kDa laminin receptor; 37LRP; 40S ribosomal protein SA; 67 kDa laminin receptor; 67LR; Colon carcinoma laminin-binding protein; LAMBR; Laminin receptor 1; laminin receptor 1 (67kD, ribosomal protein SA); Laminin-binding protein precursor p40; LamR; LAMR1; LBP/p40; LRP; LRP/LR; Multidrug resistance-associated protein MGr1-Ag; NEM/1CHD4; p40; Small ribosomal subunit protein uS2
Gene Aliases: 37LRP; 67LR; ICAS; LAMBR; lamR; LAMR1; LBP; LBP/p40; LRP; LRP/LR; NEM/1CHD4; p40; RPSA; SA
UniProt ID: (Human) P08865
Entrez Gene ID: (Human) 3921
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.