Invitrogen
Manufacturer Code:PA529743
Catalog # PIPA529743
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 55 and 284 of Human RPSA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 37 kDa laminin receptor; 37 kDa laminin receptor precursor; 37 kDa oncofetal antigen; 37/67 kDa laminin receptor; 37LRP; 37LRP 67LR LAMBR LAMR1 LBP LBP/p40 LRP LRP/LR NEM/1CHD4 SA lamR p40 37 kDa laminin receptor 37/67 kDa laminin receptor 40S ribosomal protein SA 67 kDa laminin receptor 67kD ribosomal protein SA colon carcinoma laminin-binding protein laminin binding protein laminin receptor 1 (67kD ribosomal protein SA) laminin-binding; 40S ribosomal protein SA; 67 kDa laminin receptor; 67LR; Colon carcinoma laminin-binding protein; Laminin receptor 1; laminin receptor 1 (67kD, ribosomal protein SA); Laminin-binding protein precursor p40; LamR; LBP/p40; LRP/LR; Multidrug resistance-associated protein MGr1-Ag; NEM/1CHD4; OFA/iLRP; P40-3, functional; P40-8, functional; Small ribosomal subunit protein uS2
Gene Aliases: 37LRP; 67kDa; 67LR; AL022858; ICAS; LAMBR; Lamr; LAMR1; Lamrl1; LBP; LBP/p40; LRP; LRP/LR; MLR; NEM/1CHD4; P40; P40-3; P40-8; RPSA; SA
UniProt ID: (Human) P08865, (Mouse) P14206
Entrez Gene ID: (Human) 3921, (Mouse) 16785
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.