Invitrogen
Manufacturer Code:PA5115854
Catalog # PIPA5115854
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RPS9 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3010033P07Rik 40S ribosomal protein S9 AL022771 AL022885 ribosomal protein S9 Rps9 RS9 S9 Small ribosomal subunit protein uS4; 40S ribosomal protein S9; Small ribosomal subunit protein uS4
Gene Aliases: 3010033P07Rik; AL022771; AL022885; RPS9; S9
UniProt ID: (Human) P46781, (Mouse) Q6ZWN5, (Rat) P29314
Entrez Gene ID: (Human) 6203, (Mouse) 76846, (Rat) 81772
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.