Proteintech
Manufacturer Code:172961AP150UL
Catalog # 50-173-1496
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPS4Y1 Fusion Protein Ag11176 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40S ribosomal protein S4, X isoform; 40S ribosomal protein S4, Y; 40S ribosomal protein S4, Y isoform 1; cell cycle gene 2; ribosomal protein S4, X-linked X-like; ribosomal protein S4X isoform; ribosomal protein S4Y; RPS4Y RPS4Y1; SCR10; Single copy abundant mRNA protein; single-copy abundant mRNA; Small ribosomal subunit protein eS4
Gene Aliases: CCG2; DXS306; PRO2646; RPS4; Rps4-1; RPS4X; RPS4Y; RPS4Y1; S4; SCAR; SCR10
UniProt ID: (Rat) P62703, (Human) P62701, (Human) P22090, (Mouse) P62702
Entrez Gene ID: (Rat) 100362640, (Human) 6191, (Human) 6192, (Mouse) 20102
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.