Proteintech
Manufacturer Code:141231AP150UL
Catalog # 50-172-9227
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPS3A Fusion Protein Ag5273 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40S ribosomal protein S3a; 40S ribosomal protein S3a Fte 1 FTE1 MFTL ribosomal protein S3A RPS3A; 40S ribosomal protein S3a {ECO:0000255|HAMAP-Rule:MF_03122}; 40S ribosomal protein S3b; Fte-1; Protein TU-11; ribosomal protein S3A; RPS3A; Small ribosomal subunit protein eS1; V-fos transformation effector protein; v-fos transformation effector protein 1
Gene Aliases: Fte-1; FTE1; MFTL; RPS3A; Rps3a1; S3A
UniProt ID: (Human) P61247, (Rat) P49242, (Mouse) P97351
Entrez Gene ID: (Human) 6189, (Rat) 29288, (Mouse) 20091
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.