Novus Biologicals
Manufacturer Code:NBP298931JF549
Catalog # NB056610
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human RPS3. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40S ribosomal protein S3; 40S ribosomal protein S3 FLJ26283 FLJ27450 IMR-90 ribosomal protein S3 MGC87870 ribosomal protein S3; IMR-90 ribosomal protein S3; Small ribosomal subunit protein uS3
Gene Aliases: OK/SW-cl.26; RPS3; S3
UniProt ID: (Human) P23396
Entrez Gene ID: (Human) 6188
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.