Proteintech
Manufacturer Code:119901AP150UL
Catalog # 50-560-143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPS3 Fusion Protein Ag3918 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40S ribosomal protein S3; 40S ribosomal protein S3 ribosomal protein S3 RPS3; IMR-90 ribosomal protein S3; Small ribosomal subunit protein uS3
Gene Aliases: D7Ertd795e; OK/SW-cl.26; RPS3; Rs_3; S3
UniProt ID: (Human) P23396, (Mouse) P62908
Entrez Gene ID: (Human) 6188, (Mouse) 27050
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.