Invitrogen
Manufacturer Code:PA544506
Catalog # PIPA544506
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human RPS21 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40S ribosomal protein S21; 8.2 kDa differentiation factor; human leukemia differentiation factor; Ribosomal protein S21 S21; Small ribosomal subunit protein eS21
Gene Aliases: 1810049N11Rik; 2410030A14Rik; HLDF; RPS21; S21; zgc:56642; zgc:86816
UniProt ID: (Bovine) Q32PB8, (Human) P63220, (Mouse) Q9CQR2, (Rat) P05765, (Zebrafish) Q7ZUG5
Entrez Gene ID: (Bovine) 615178, (Rabbit) 100342677, (Human) 6227, (Dog) 485959, (Mouse) 66481, (Guinea pig) 100735882, (Rat) 81775, (Zebrafish) 394166
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.