Invitrogen
Manufacturer Code:PA5103534
Catalog # PIPA5103534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RPS21 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810049N11Rik 2410030A14Rik 40S ribosomal protein S21 8.2 kDa differentiation factor HLDF human leukemia differentiation factor ribosomal protein S21 RPS21 S21 Small ribosomal subunit protein eS21; 40S ribosomal protein S21; 8.2 kDa differentiation factor; human leukemia differentiation factor; Small ribosomal subunit protein eS21
Gene Aliases: 1810049N11Rik; 2410030A14Rik; HLDF; RPS21; S21
UniProt ID: (Human) P63220, (Mouse) Q9CQR2, (Rat) P05765
Entrez Gene ID: (Human) 6227, (Mouse) 66481, (Rat) 81775
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.