Invitrogen
Manufacturer Code:PA572341
Catalog # PIPA572341
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 45-79 amino acids from human RPS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40S ribosomal protein S2; 40S ribosomal protein S4; LLREP3 S2 40S ribosomal protein S2 40S ribosomal protein S4 OK/KNS-cl.6 protein LLRep3; OK/KNS-cl.6; Protein LLRep3; repeat family 3; Small ribosomal subunit protein uS5
Gene Aliases: LLREP3; RPS2; RPS4; S2; S4
UniProt ID: (Human) P15880, (Mouse) P25444
Entrez Gene ID: (Human) 6187, (Mouse) 16898
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.