Novus Biologicals
Manufacturer Code:NBP214853
Catalog # NBP214853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody maps to a region between residue 243 and 293 of human Ribosomal Protein S2 using the numbering given in entry NP_002943.2 (GeneID 6187). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40S ribosomal protein S2; 40S ribosomal protein S4; LLREP3 MGC102851 MGC117344 MGC11734540S ribosomal protein S4 OK/KNS-cl.640S ribosomal protein S2 Protein LLRep3 ribosomal protein S2 RPS4; OK/KNS-cl.6; Protein LLRep3; Small ribosomal subunit protein uS5
Gene Aliases: LLREP3; RPS2; RPS4; S2
UniProt ID: (Human) P15880
Entrez Gene ID: (Human) 6187
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.