Abnova
Manufacturer Code:H00006187A01
Catalog # 89-105-360
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 198 a.a. &bsim 293 a.a) partial recombinant protein with GST tag.,RPS2 (NP_002943 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant RPS2.
For Research Use Only
Protein Aliases: 40S ribosomal protein S2; 40S ribosomal protein S4; LLREP3; MGC102851; MGC117344; MGC117345; OK/KNS-cl.6; Protein LLRep3; Small ribosomal subunit protein uS5
Gene Aliases: LLREP3; RPS2; RPS4; S2
UniProt ID: (Human) P15880
Entrez Gene ID: (Human) 6187
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.