Invitrogen
Manufacturer Code:MA535388
Catalog # PIMA535388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0820 |
Immunogen | A synthesized peptide derived from human RPS19 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 40S ribosomal protein S19; 40S ribosomal protein S19 DBA DBA1 Dsk3 HGNC:10402 Rps19 S19 Small ribosomal subunit protein eS19; Small ribosomal subunit protein eS19
Gene Aliases: DBA; DBA1; Dsk3; RPS19; S19
UniProt ID: (Human) P39019, (Rat) P17074, (Mouse) Q9CZX8
Entrez Gene ID: (Human) 6223, (Rat) 29287, (Mouse) 20085
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.