Invitrogen
Manufacturer Code:PA518452
Catalog # PIPA518452
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KMVEKDQDGGRK) corresponding to the internal amino acids of RPS19 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 40S ribosomal protein S19; RPS19 ribosomal protein S19 HGNC:10402 DBA 40S ribosomal protein S19; Small ribosomal subunit protein eS19
Gene Aliases: DBA; DBA1; RPS19; S19
UniProt ID: (Human) P39019
Entrez Gene ID: (Human) 6223
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.