Proteintech
Manufacturer Code:166801AP150UL
Catalog # 50-173-1108
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPS13 Fusion Protein Ag10008 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40S ribosomal protein S13; 40S ribosomal protein S13 ribosomal protein S13 RPS13; Small ribosomal subunit protein uS15
Gene Aliases: 2700063M04Rik; RPS13; S13
UniProt ID: (Human) P62277, (Rat) P62278, (Mouse) P62301
Entrez Gene ID: (Human) 6207, (Rat) 161477, (Mouse) 68052
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.