OriGene
Manufacturer Code:CF507369
Catalog # 50-166-4755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C9 |
Immunogen | Full length human recombit protein of human RPRD1B produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C20orf77 CREPT K-H Kub5-Hera NET60 dJ1057B20.2; Cell cycle-related and expression-elevated protein in tumor; cell-cycle related and expression-elevated protein in tumor; Regulation of nuclear pre-mRNA domain-containing protein 1B
Gene Aliases: C20orf77; CREPT; dJ1057B20.2; NET60; RPRD1B
UniProt ID: (Human) Q9NQG5
Entrez Gene ID: (Human) 58490
Molecular Function:
enzyme modulator
kinase inhibitor
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.