Proteintech
Manufacturer Code:128741AP150UL
Catalog # 50-172-8318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPP38 Fusion Protein Ag3552 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ribonuclease P protein subunit p38; ribonuclease P/MRP 38kDa subunit; RNaseP protein p38; RNaseP protein p38 RP11 455B2.5 RPP38
Gene Aliases: C330006A15Rik; RPP38
UniProt ID: (Human) P78345
Entrez Gene ID: (Human) 10557, (Mouse) 227522, (Rat) 291317
Molecular Function: RNA binding protein endoribonuclease esterase hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.