Invitrogen
Manufacturer Code:PA558019
Catalog # PIPA558019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RPP30 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ribonuclease P protein subunit p30; ribonuclease P/MRP 30kDa subunit; RNase P subunit 2; RNaseP protein p30; TSG15 RNase P subunit 2 RNaseP protein p30 ribonuclease P (30kD) (RPP30) ribonuclease P protein subunit p30
Gene Aliases: RNASEP2; RPP30; TSG15
UniProt ID: (Human) P78346
Entrez Gene ID: (Human) 10556
Molecular Function: RNA binding protein endoribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.