Novus Biologicals
Manufacturer Code:NBP294695100UL
Catalog # NB023914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-268 of human RPP30 (NP_006404.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.26.5 FLJ38491 ribonuclease P (30kD) (RPP30) ribonuclease P protein subunit p30 ribonuclease P/MRP 30kDa subunit RNase P subunit 2 RNaseP protein p30 RNASEP2 TSG15; Ribonuclease P protein subunit p30; ribonuclease P/MRP 30kDa subunit; RNase P subunit 2; RNaseP protein p30
Gene Aliases: RNASEP2; RPP30; TSG15
UniProt ID: (Human) P78346
Entrez Gene ID: (Human) 10556
Molecular Function: RNA binding protein endoribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.