Proteintech
Manufacturer Code:154611AP150UL
Catalog # 50-173-0247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPP25 Fusion Protein Ag7742 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ribonuclease P 25 subunit; ribonuclease P 25kDa subunit; Ribonuclease P protein subunit p25; ribonuclease P/MRP 25kDa subunit; RNase P protein subunit p25; RNase P protein subunit p25 RPP25
Gene Aliases: AI851155; RPP25
UniProt ID: (Human) Q9BUL9, (Rat) Q5PPN2, (Mouse) Q91WE3
Entrez Gene ID: (Human) 54913, (Rat) 315705, (Mouse) 102614
Molecular Function:
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.