Invitrogen
Manufacturer Code:PA5113486
Catalog # PIPA5113486
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Ribonuclease P protein subunit p21 protein (2-154AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610037N01Rik C6orf135 Cat60 Flj22638 ribonuclease P 21 subunit ribonuclease P 21kDa subunit Ribonuclease P protein subunit p21 Ribonuclease P/MRP 21 kDa subunit ribonuclease P/MRP 21 subunit ribonuclease P/MRP 21kDa subunit ribonuclease P/MRP subunit p21 Ribonucleoprotein V RNaseP protein p21 Rpp21; Ribonuclease P protein subunit p21; Ribonuclease P/MRP 21 kDa subunit; ribonuclease P/MRP 21kDa subunit; Ribonucleoprotein V; RNaseP protein p21
Gene Aliases: C6orf135; CAT60; RPP21
UniProt ID: (Human) Q9H633
Entrez Gene ID: (Human) 79897
Molecular Function: hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.