Novus Biologicals
Manufacturer Code:NBP2842530.1ML
Catalog # NB032181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of RPP21 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf135 CAT60 chromosome 6 open reading frame 135 EC 3.1.26.5 Em:AB014085.3 FLJ22638 ribonuclease P 21kDa subunit ribonuclease P protein subunit p21 Ribonuclease P/MRP 21 kDa subunit ribonuclease P/MRP 21kDa subunit Ribonucleoprotein V RNaseP protein p21; Ribonuclease P protein subunit p21; Ribonuclease P/MRP 21 kDa subunit; ribonuclease P/MRP 21kDa subunit; Ribonucleoprotein V; RNaseP protein p21
Gene Aliases: C6orf135; CAT60; RPP21
UniProt ID: (Human) Q9H633
Entrez Gene ID: (Human) 79897
Molecular Function: hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.