Proteintech
Manufacturer Code:163771AP150UL
Catalog # 50-173-0905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPP21 Fusion Protein Ag9353 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf135 CAT60 Ribonucleoprotein V RNaseP protein p21 RPP21; ribonuclease P 21 subunit; ribonuclease P 21kDa subunit; Ribonuclease P protein subunit p21; Ribonuclease P/MRP 21 kDa subunit; ribonuclease P/MRP 21kDa subunit; Ribonucleoprotein V; RNaseP protein p21
Gene Aliases: 0610037N01Rik; C6orf135; CAT60; Flj22638; RPP21
UniProt ID: (Human) Q9H633, (Mouse) Q8R040
Entrez Gene ID: (Human) 79897, (Mouse) 67676, (Rat) 406230
Molecular Function: hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.