Invitrogen
Manufacturer Code:PA5100323
Catalog # PIPA5100323
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RPN1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU018702 D6Wsu137e Dolichyl-diphosphooligosaccharide--protein dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1 oligosaccharyltransfe oligosaccharyltransferase 1 homolog oligosaccharyltransferase complex subunit (non-catalytic) OST1 RBPH1 RIBI Ribophorin I ribophorin-1 RPN1 Rpn-1 RPN-I; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1; dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit; oligosaccharyltransferase 1 homolog; oligosaccharyltransferase complex subunit (non-catalytic); Ribophorin I; Ribophorin-1; RPN-I
Gene Aliases: AU018702; D6Wsu137e; OST1; RBPH1; RIBI; Rpn-1; RPN-I; RPN1
UniProt ID: (Human) P04843, (Mouse) Q91YQ5
Entrez Gene ID: (Human) 6184, (Rat) 25596, (Mouse) 103963
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.