Novus Biologicals
Manufacturer Code:NBP294116100UL
Catalog # NB022789
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-114 of human RPLP1 (NP_000994.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60S acidic ribosomal protein P1; acidic ribosomal phosphoprotein P1; acidic ribosomal phosphoprotein P1 FLJ27448 MGC5215 P160S acidic ribosomal protein P1 ribosomal protein large P1 RPP1 RRP1; Large ribosomal subunit protein P1; ribosomal protein, large, P1
Gene Aliases: LP1; P1; RPLP1; RPP1; RRP1
UniProt ID: (Human) P05386
Entrez Gene ID: (Human) 6176
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.