Novus Biologicals
Manufacturer Code:NBP220220
Catalog # NBP220220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human RPLP0. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60S acidic ribosomal protein P0; 60S acidic ribosomal protein P0 60S ribosomal protein L10E acidic ribosomal phosphoprotein P0 L10E MGC111226 MGC88175 P0 PRLP0 ribosomal protein large P0 RPP0; 60S ribosomal protein L10E; acidic ribosomal phosphoprotein P0; Large ribosomal subunit protein uL10; neutral ribosomal phosphoprotein P0; ribosomal protein, large, P0
Gene Aliases: L10E; LP0; P0; PRLP0; RPLP0; RPP0
UniProt ID: (Human) P05388
Entrez Gene ID: (Human) 6175
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.