Invitrogen
Manufacturer Code:PA568074
Catalog # PIPA568074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human RPL38. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60S ribosomal protein L38; Large ribosomal subunit protein eL38
Gene Aliases: 0610025G13Rik; L38; Rbt; RPL38; Ts; Tss
UniProt ID: (Human) P63173, (Rat) P63174, (Mouse) Q9JJI8
Entrez Gene ID: (Human) 6169, (Rat) 689284, (Mouse) 67671
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.