Invitrogen
Manufacturer Code:PA5112770
Catalog # PIPA5112770
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human 60S ribosomal protein L38 protein (1-70AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610025G13Rik 60S ribosomal protein L38 L38 Large ribosomal subunit protein eL38 Rbt ribosomal protein L38 Rpl38 Ts Tss; 60S ribosomal protein L38; Large ribosomal subunit protein eL38
Gene Aliases: L38; RPL38
UniProt ID: (Human) P63173
Entrez Gene ID: (Human) 6169
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.