Proteintech
Manufacturer Code:148261AP150UL
Catalog # 50-172-9756
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPL35 Fusion Protein Ag6615 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60S ribosomal protein L35; 60S ribosomal protein L35 ribosomal protein L35 RPL35; Large ribosomal subunit protein uL29
Gene Aliases: 2410039E09Rik; L35; RPL35
UniProt ID: (Human) P42766, (Mouse) Q6ZWV7, (Rat) P17078
Entrez Gene ID: (Human) 11224, (Mouse) 66489, (Rat) 296709
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.