Novus Biologicals
Manufacturer Code:NBP29449320UL
Catalog # NB023515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human RPL3 (NP_000958.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60S ribosomal protein L3; 60S ribosomal protein L3 HIV-1 TAR RNA-binding protein B MGC104284 ribosomal protein L3 TARBP-B; HIV-1 TAR RNA-binding protein B; Large ribosomal subunit protein uL3; ribosomal protein L3, isoform a; TARBP-B
Gene Aliases: ASC-1; L3; OK/SW-cl.32; RPL3; TARBP-B
UniProt ID: (Human) P39023
Entrez Gene ID: (Human) 6122
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.