Abnova
Manufacturer Code:PAB20200
Catalog # 89-121-706
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human RPL3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant RPL3.
For Research Use Only
Protein Aliases: 60S ribosomal protein L3; HIV-1 TAR RNA-binding protein B; Large ribosomal subunit protein uL3; MGC104284; ribosomal protein L3, isoform a; TARBP-B
Gene Aliases: ASC-1; L3; OK/SW-cl.32; RPL3; TARBP-B
UniProt ID: (Human) P39023
Entrez Gene ID: (Human) 6122
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.