Proteintech
Manufacturer Code:110051AP150UL
Catalog # 50-172-6787
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPL3 Fusion Protein Ag1436 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60S ribosomal protein L3; 60S ribosomal protein L3 ribosomal protein L3 RPL3 TARBP B; HIV-1 TAR RNA-binding protein B; J1 protein; Large ribosomal subunit protein uL3; ribosomal protein L3 homolog; ribosomal protein L3, isoform a; TARBP-B
Gene Aliases: ASC-1; F2; J1; L3; OK/SW-cl.32; RPL3; TARBP-B
UniProt ID: (Human) P39023, (Mouse) P27659
Entrez Gene ID: (Human) 6122, (Mouse) 27367
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.