Abnova
Manufacturer Code:H00006122K
Catalog # 89-202-842
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human RPL3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human RPL3 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 60S ribosomal protein L3; HIV-1 TAR RNA-binding protein B; Large ribosomal subunit protein uL3; MGC104284 TARBP-B; ribosomal protein L3, isoform a; TARBP-B
Gene Aliases: ASC-1; L3; OK/SW-cl.32; RPL3; TARBP-B
UniProt ID: (Human) P39023
Entrez Gene ID: (Human) 6122
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.