Proteintech
Manufacturer Code:170821AP150UL
Catalog # 50-173-1374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPL24 Fusion Protein Ag7085 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60S ribosomal protein L24; 60S ribosomal protein L24 60S ribosomal protein L30 ribosomal protein L24 RPL24; 60S ribosomal protein L30; belly spot and tail; epididymis secretory protein Li 310; L30; Large ribosomal subunit protein eL24; ribosomal protein L30
Gene Aliases: 0610008L05Rik; Bst; HEL-S-310; L24; RPL24
UniProt ID: (Human) P83731, (Rat) P83732, (Mouse) Q8BP67
Entrez Gene ID: (Human) 6152, (Rat) 64307, (Mouse) 68193
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.