Invitrogen
Manufacturer Code:PA5110318
Catalog # PIPA5110318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human RPL23 (NP_000969.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810009A01Rik 60S ribosomal protein L17 60S ribosomal protein L23 L23 Large ribosomal subunit protein uL14 MGC111167 MGC117346 MGC72008 rpL17 Rpl23; 60S ribosomal protein L17; 60S ribosomal protein L23; Large ribosomal subunit protein uL14; ribosomal protein L17
Gene Aliases: 2810009A01Rik; L23; rpL17; RPL23
UniProt ID: (Human) P62829, (Mouse) P23131, (Rat) P62832
Entrez Gene ID: (Human) 9349, (Mouse) 65019, (Rat) 29282
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.