Novus Biologicals
Manufacturer Code:NBP273944FR
Catalog # NBP273944FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6B11 |
Immunogen | Full length recombinant protein of human RPL10 (NP_006004) produced in E.coli. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 60S ribosomal protein L10; 60S ribosomal protein L10 DXS648EFLJ27072 DXS648FLJ23544 Laminin receptor homolog NOV Protein QM QMDKFZp686J1851 ribosomal protein L10 Tumor suppressor QM Wilms tumor-related protein; Laminin receptor homolog; Large ribosomal subunit protein uL16; Protein QM; Ribosomal protein L10; Tumor suppressor QM; Wilms tumor-related protein
Gene Aliases: AUTSX5; DXS648; DXS648E; L10; NOV; QM; RPL10
UniProt ID: (Human) P27635
Entrez Gene ID: (Human) 6134
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.