Invitrogen
Manufacturer Code:PA538249
Catalog # PIPA538249
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human 60S Ribosomal Protein L10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 24.6 kDa protein; 24.6kDa protein; 60S ribosomal protein L10; Laminin receptor homolog; Large ribosomal subunit protein uL16; Protein QM; Protein QM homolog; QM RPLPO; ribosomal protein 10; Ribosomal protein L10; Tumor suppressor QM; Wilms tumor-related protein
Gene Aliases: AUTSX5; D0HXS648; DXHXS648; DXHXS648E; DXS648; DXS648E; L10; NOV; QM; QM16; RPL10
UniProt ID: (Human) P27635, (Rat) Q6PDV7, (Mouse) Q6ZWV3
Entrez Gene ID: (Human) 6134, (Rat) 81764, (Mouse) 110954
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.