Invitrogen
Manufacturer Code:PA5101098
Catalog # PIPA5101098
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RL10 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 24.6 kDa protein; 24.6 kDa protein 24.6kDa protein 60S ribosomal protein L10 AUTSX5 D0HXS648 DXHXS648 DXHXS648E DXS648 DXS648E L10 Laminin receptor homolog Large ribosomal subunit protein uL16 NOV Protein QM Protein QM homolog QM QM16 ribosomal protein 10 ribosomal protein L10 RPL10 RPLPO tumor suppressor QM Wilms tumor-related protein; 24.6kDa protein; 60S ribosomal protein L10; Laminin receptor homolog; Large ribosomal subunit protein uL16; Protein QM; Protein QM homolog; ribosomal protein 10; Ribosomal protein L10; Tumor suppressor QM; Wilms tumor-related protein
Gene Aliases: AUTSX5; D0HXS648; DXHXS648; DXHXS648E; DXS648; DXS648E; L10; NOV; QM; QM16; RPL10
UniProt ID: (Human) P27635, (Rat) Q6PDV7, (Mouse) Q6ZWV3
Entrez Gene ID: (Human) 6134, (Rat) 81764, (Mouse) 110954
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.