Abnova
Manufacturer Code:H00006134K
Catalog # 89-202-848
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human RPL10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human RPL10 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 60S ribosomal protein L10; DKFZp686J1851 DXS648 DXS648E FLJ23544 FLJ27072 NOV QM; Laminin receptor homolog; Large ribosomal subunit protein uL16; Protein QM; Ribosomal protein L10; Tumor suppressor QM; Wilms tumor-related protein
Gene Aliases: AUTSX5; DXS648; DXS648E; L10; NOV; QM; RPL10
UniProt ID: (Human) P27635
Entrez Gene ID: (Human) 6134
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.