Novus Biologicals
Manufacturer Code:NBP273943IR
Catalog # NBP273943IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3C6 |
Immunogen | Full length recombinant protein of human RPIP8 (NP_006686) produced in HEK293T cells. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: RaP2 interacting protein 8; Rap2-interacting protein 8; Rap2-interacting protein 8 RAP2IPRUN domain-containing protein 3A RPIP-8 RPIP8RaP2 interacting protein 8 RUN domain containing 3A; RPIP-8; RUN domain-containing protein 3A
Gene Aliases: RAP2IP; RPIP-8; RPIP8; RUNDC3A
UniProt ID: (Human) Q59EK9
Entrez Gene ID: (Human) 10900
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.