Novus Biologicals
Manufacturer Code:NBP220209
Catalog # NBP220209
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human RPIP8. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RaP2 interacting protein 8; Rap2-interacting protein 8; Rap2-interacting protein 8 RAP2IPRUN domain-containing protein 3A RPIP-8 RPIP8RaP2 interacting protein 8 RUN domain containing 3A; RPIP-8; RUN domain-containing protein 3A
Gene Aliases: RAP2IP; RPIP-8; RPIP8; RUNDC3A
UniProt ID: (Human) Q59EK9
Entrez Gene ID: (Human) 10900
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.