Invitrogen
Manufacturer Code:PA5112699
Catalog # PIPA5112699
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Ribose-5-phosphate isomerase protein (82-311AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOS_11661 LOC100347639 Phosphoriboisomerase ribose 5-phosphate epimerase ribose 5-phosphate isomerase A ribose 5-phosphate isomerase A (ribose 5-phosphate epimerase) Ribose-5-phosphate isomerase RPI RPIA RPIAD; Phosphoriboisomerase; ribose 5-phosphate epimerase; Ribose-5-phosphate isomerase
Gene Aliases: RPI; RPIA; RPIAD
UniProt ID: (Human) P49247, (Mouse) P47968
Entrez Gene ID: (Human) 22934, (Mouse) 19895
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.