Proteintech
Manufacturer Code:152971AP150UL
Catalog # 50-173-0130
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPH3AL Fusion Protein Ag7552 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: No C2 domains protein; No C2 domains protein NOC2 Rab effector Noc2 Rabphilin 3A like protein RPH3AL; Rab effector Noc2; Rabphilin-3A-like protein
Gene Aliases: NOC2; RPH3AL
UniProt ID: (Human) Q9UNE2
Entrez Gene ID: (Human) 9501
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.